Haz-Dust Real-Time Dust Monitor
Provides instant airborne particulate concentrations in accordance with NIOSH method #0600
  • Measures concentration levels from 0.01 up to 200 mg/m3
  • Ideal for industrial hygiene, indoor air quality, and haz-mat monitoring
Affordable, hand-held survey monitor is lightweight and easy to operate. Simply turn monitor on and immediately view dust concentrations on digital display. Sensing range is easily selected from 0.1 to 200 or 0.01 to 20 mg/m3 at the flip of a switch . Haz-Dust monitor is appropriate for a variety of aerosols and dusts including: lead, silica, coal, wood, dry chemicals, welding fumes, smoke, paint spray, and cadmium.
Rugged, well-constructed ABS plastic case protects monitor electronics from accidental drops. Unique design provides protection against RFI/EMI waves.
What's included: rechargeable battery with 110/230 VAC, 50/60 Hz charger and carrying case.
£4,579.10GBP / Each
£5,494.92GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max CO2 (ppm) Max Humidity (RH) Max Oxygen (%)
Availability Pricing
Environmental Devices HD-1100 Environemental Devices Haz-Dust Real-Time Dust Monitor WZ-86553-00 Mfr # HD-1100
COMPARE
£4,579.10GBP / Each
£5,494.92GBP (incl VAT)