Hayward® True Union Ball Check Valves with Foot Valve Screens
Unique square-cut elastomer seat provides seal at very low back pressures
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.
The foot valve screen helps prevent unwanted material from entering the system.
£57.75 - £87.15GBP / Each
£69.30 - £104.58GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Cv Port Size (NPT(F))
Availability Pricing
Hayward FV10050SA Foot valve screen for ball valve 01341-00, 1/2" WZ-01341-50 Mfr # FV10050SA
COMPARE
4.6 0.5
£57.75GBP / Each
£69.30GBP (incl VAT)
Hayward FV10075SA Foot valve screen for ball valve 01341-02, 3/4" WZ-01341-52 Mfr # FV10075SA
COMPARE
10 0.75
£59.85GBP / Each
£71.82GBP (incl VAT)
Hayward FV10150SA Foot valve screen for ball valve 01341-08, 1-1/2" WZ-01341-58 Mfr # FV10150SA
COMPARE
55 1.5
£87.15GBP / Each
£104.58GBP (incl VAT)