This item is now discontinued. Please browse similar items
Hach EC5 5061 SensION+ Rugged Conductivity Meter Kit
Representative image only.
Hach Mfr # LPV3562.97.0002 – Item # WZ-19604-03
All-in-one kits with everything you need to start testing
  • Guided, menu navigation makes conductivity testing fast and simple
  • Waterproof, IP67-rated housing
  • Large backlit LCD with anti-glare coating
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypePortable
  • Display typeLCD
  • Min Conductivity0.01 µS to 500 mS
  • Conductivity Resolution0.01, 0.1, or 1 μS; 0.01, 0.1, or 1 mS
  • Conductivity Accuracy±0.5%
  • TDS Accuracy±0.5%
  • Salinity Resolution0.1 or 1 ppm
  • Temperature Accuracy±0.3°C
  • Temperature compensationAutomatic or manual
  • Battery3 x AA batteries (included)
  • Battery life (hr)500
  • DescriptionSensION+ Rugged Conductivity Meter Kit
  • Warranty3 year
3 Year Warranty
Key Features
  • Guided, menu navigation makes conductivity testing fast and simple
  • Waterproof, IP67-rated housing
  • Large backlit LCD with anti-glare coating
  • 3 year warranty
More About this Item
The sensION EC5 conductivity, TDS, salinity and temperature meter is packaged with everything you need to start testing for general aqueous or difficult media applications. Featuring a workstation carrying case, 3-point calibration, temperature re-adjustment feature and improved stabilization algorithms.
General aqueous EC5 kit includes conductivity probe 19604-51.
Advanced EC5 kit includes robust titanium conductivity probe 19604-52.