H-B Instrument Metal Thermometer Case
Brass armor protects glass thermometer from damage
Nickel plated brass armor case is designed to protect liquid-in-glass thermometers from accidental breakage during use and storage. The case is available in sizes ranging from 254 mm to 610 mm and features a screw cap with ring and small openings around bulb for circulation.
£31.78 - £52.58GBP / Each
£38.14 - £63.10GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
H-B Instrument B61400-9100 Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 254mm WZ-90311-14 Mfr # 614009100
COMPARE
Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 254mm
£31.78GBP / Each
£38.14GBP (incl VAT)
H-B Instrument B61400-9400 Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 380mm WZ-90311-15 Mfr # 614009400
COMPARE
Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 380mm
£44.99GBP / Each
£53.99GBP (incl VAT)
H-B Instrument B61400-9500 Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 405mm WZ-90311-16 Mfr # 614009500
COMPARE
Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 405mm
£44.99GBP / Each
£53.99GBP (incl VAT)
H-B Instrument B61400-9600 Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 460mm WZ-90311-17 Mfr # 614009600
COMPARE
Nickel Plated Brass Armor Liquid-in-Glass Thermometer Case, 460mm
£52.58GBP / Each
£63.10GBP (incl VAT)