This product is now discontinued, View Related Products
H-B Instrument Glass Thermometer Cases
Keep glass thermometers safe during transport or while not in use
  • Properly protect all glass thermometers
To guard against separation, store thermometers horizontally. The plush lining protects thermometers against breakage.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
H-B Instrument B61400-3100 Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 300mm WZ-08118-72 Mfr # B61400-3100
COMPARE
Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 300mm
Call For Price
H-B Instrument B61400-3200 Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 405mm WZ-08118-76 Mfr # B61400-3200
COMPARE
Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 405mm
Call For Price
H-B Instrument B61400-3300 Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 460mm WZ-08118-78 Mfr # B61400-3300
COMPARE
Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 460mm
Call For Price
H-B Instrument B61400-3400 Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 610mm WZ-08118-84 Mfr # B61400-3400
COMPARE
Velvet Lined Leatherette Liquid-in-Glass Thermometer Storage Case; Single, 610mm