This product is now discontinued, View Related Products
H-B Instrument Easy-Read Enviro-Safe Nonhazardous Pocket Glass Thermometers, Fahrenheit-scale
Rugged case with an eyelet at the top for easy suspension
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in two case styles; closed metal, or open metal. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.
Easy-Read® thermometers feature a black liquid and yellow back glass - distinct contrast makes these thermometers very easy to read.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
H-B Instrument B60370-0000 Easy-Read Pocket Liquid-In-Glass Thermometer; 0 to 220F, Closed Metal Case, Environmentally Friendly WZ-90300-36 Mfr # B60370-0000
COMPARE
220 0 160
Call For Price
H-B Instrument B60370-0300 Easy-Read Pocket Liquid-In-Glass Thermometer; 20 to 120F, Closed Metal Case, Environmentally Friendly WZ-90300-39 Mfr # B60370-0300
COMPARE
120 20 160
Call For Price
H-B Instrument B60370-0500 Easy-Read Pocket Liquid-In-Glass Thermometer; 0 to 220F, Window Metal Case, Environmentally Friendly WZ-90300-42 Mfr # B60370-0500
COMPARE
220 0 160
Call For Price
H-B Instrument B60370-0800 Easy-Read Pocket Liquid-In-Glass Thermometer; 20 to 120F, Window Metal Case, Environmentally Friendly WZ-90300-45 Mfr # B60370-0800
COMPARE
120 20 160
Call For Price
H-B Instrument B60370-1000 Easy-Read Pocket Liquid-In-Glass Thermometer; 0 to 220F, Window Plastic Case, Environmentally Friendly WZ-90300-47 Mfr # B60370-1000
COMPARE
220 0 160