Cole-Parmer Durac Polycarbonate Brix Sugar Hydrometers, plain form
These durable polycarbonate hydrometers won't break when dropped—perfect for industrial or laboratory use.
The temperature standard for these hydrometers is 60°F (15°C). All hydrometers are individually serialized, feature metal ballast with wax epoxy and include multi-language statement of accuracy supplied by the vendor.
Available in two models measuring sugar from 0° to 35° or 15° to 50° Brix, these hydrometers measure % sucrose by weight.
Note: Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
£132.55GBP / Each
£159.06GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Brix Scale Range (Max) (%) Brix Scale Range (Min) (%) Hydrometer Form
Availability Pricing
COMPARE
Plain
£132.55GBP / Each
£159.06GBP (incl VAT)