BriskHeat® Grounded Heating Tapes - 230 VAC
Safe for heating metal vessels
  • Heat to 482F (250C)
  • NOTE: Temperature range has been reduced to meet safty standards
Wind heating tape around glass, ceramic, or metal vessels—grounding prevents shock hazard. Temperature-resistant fiberglass sleeves encase heating element; ground wire at core. The maximum surface loading is 4 watts/sq in. Leads are 31/2" long with electrical connector at one end. Tapes include ties at both ends.
WARNING: Heating tape should not be overlapped and require the use of a temperature or power controller sold separately.
£104.57 - £425.86GBP / Each
£125.48 - £511.03GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat CPBIH102020LGT Grounded Fiberglass Insulated Heat Tape, 2' x 1" , 100 Watts, 230 V WZ-03121-12 Mfr # CPBIH102020LGT
COMPARE
2 482 230
£104.57GBP / Each
£125.48GBP (incl VAT)
BriskHeat CPBIH102040LGT Grounded Fiberglass Insulated Heat Tape, 4' x 1" , 200 Watts, 230 V WZ-03121-32 Mfr # CPBIH102040LGT
COMPARE
4 482 230
£123.23GBP / Each
£147.88GBP (incl VAT)
BriskHeat CPBIH102060LGT Grounded Fiberglass Insulated Heat Tape, 6' x 1" , 300 Watts, 230 V WZ-03121-42 Mfr # CPBIH102060LGT
COMPARE
6 482 230
£152.30GBP / Each
£182.76GBP (incl VAT)
BriskHeat CPBIH102032LGT Grounded Fiberglass Insulated Heat Tape, 32' x 1" , 1600 Watts, 230 V WZ-03121-92 Mfr # CPBIH102032LGT
COMPARE
32 482 230
£425.86GBP / Each
£511.03GBP (incl VAT)