Cole-Parmer® Graphite Capillary Column Ferrules
Reusable one-piece design
  • Constructed of durable graphite
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450°C. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.
£51.03 - £52.50GBP / Pkg of 10
£61.24 - £63.00GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Graphite Capillary Column Ferrules, 1/16\" to 0.4 mm
£52.50GBP / Pkg of 10
£63.00GBP (incl VAT)
COMPARE
Graphite Capillary Column Ferrules, 1/16\" to 0.5 mm
£51.03GBP / Pkg of 10
£61.24GBP (incl VAT)
COMPARE
Graphite Capillary Column Ferrules, 1/16\" to 0.8 mm
£52.50GBP / Pkg of 10
£63.00GBP (incl VAT)