This product is now discontinued, View Related Products
Cole-Parmer® essentials Graduated Sample Containers, PP
No more leaks--leakproof for better sampling
  • Easier sampling--graduated for measuring directly
  • Durable design--translucent, unbreakable, and chemical resistant
Graduated polypropylene (PP) containers are translucent, unbreakable, and chemical-resistant. Containers tolerate strong acids, alkalies, and most organic solvents. Full-turn screw thread provides leakproof seal. Choose from sterile or nonsterile versions; sterile containers are individually bagged. Containers come with identification labels and color-coded HDPE caps (green-sterile; white-nonsterile).
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (oz) Overall Height (in) Special Features
Availability Pricing
COMPARE
2 2 5/8
Call For Price
COMPARE
4 3