Cole-Parmer® Essentials Graduated Bottles, Clear Glass
Less spilling during transfer
These sealable graduated glass bottles are ideal for samples that need to be monitored or measured as they are used. Each is made of clear, undistorted, USP Type III soda-lime glass and graduated in both milliliters and ounces. Wide-mouth design reduces the risk of spillage during transfer, while thermoset screw-on caps ensure samples remain unaffected by airborne contaminants like gas, oil, or chemical fumes. Special, 32-oz beaker utilizes convenient, French square design, allowing a more economical use of shelf space while making the container easier to grip when pouring.
£191.67 - £272.68GBP / Case of 48
£230.00 - £327.22GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Cole-Parmer Essentials 7981 Graduated Round Bottle, Clear Glass, 30mL; 48/CS WZ-34601-10 Mfr # GLC-01450
COMPARE
30 5 Glass
£226.27GBP / Case of 48
£271.52GBP (incl VAT)
Cole-Parmer Essentials 7982 Graduated Round Bottle, Clear Glass, 60mL; 48/CS WZ-34601-20 Mfr # GLC-01472
COMPARE
60 5 Glass
£272.68GBP / Case of 48
£327.22GBP (incl VAT)
Cole-Parmer Essentials 7983 Graduated Round Bottle, Clear Glass, 120mL; 24/CS WZ-34601-30 Mfr # GLC-01504
COMPARE
125 5 Glass
£191.67GBP / Case of 24
£230.00GBP (incl VAT)
Cole-Parmer Essentials 7984 Graduated Round Bottle, Clear Glass, 250mL; 24/CS WZ-34601-40 Mfr # GLC-01532
COMPARE
250 20 Glass
£247.54GBP / Case of 24
£297.05GBP (incl VAT)
Cole-Parmer Essentials 7907 Graduated Square Bottle, Clear Glass, 1000mL; 12/CS WZ-34601-60 Mfr # GLC-01388
COMPARE
1000 25 Glass
£233.36GBP / Case of 12
£280.03GBP (incl VAT)