This product is now discontinued, View Related Products
Corning® Gosselin™ Traceable Standard-Sized Petri Dishes, PS
Culture dishes for any application
  • Injection point located near the edge of the bottom maintains optimal clarity for imaging systems Lightweight polystyrene (PS) is compliant with heavy metals (94/62/EC) and food contact (EC 1935/2004) directives Class 100 manufacturing environment ensures aseptic production and packaging Vented for easy gas exchange
Traceable standard petri dishes are available in nonsterile and sterile (SAL 10⁻⁶) versions, crystal or optically clear polystyrene, and feature cleanroom packaging with two outer bags and bar code label.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Sterile
Availability Pricing
Corning Gosselin Sterile Petri Dish w/ Frosted Area, double bagged, traceability labels; 825/pk WZ-34551-85 Mfr # BP93D-102
COMPARE
Crystal polystyrene w/ frosted area Yes
Call For Price
Corning Gosselin Sterile Petri Dish, 60 x 15 mm, double bagged, traceability labels; 1620/pk WZ-34552-07 Mfr # BP53-03
COMPARE
Crystal polystyrene Yes