This product is now discontinued, View Related Products

Corning® Gosselin™ Straight-Sided Screw-Cap Containers, PP

Safely collect and transport media samples
  • Keep samples safe inside a shockproof PP container
  • Screw-cap closure with inner lip for a leakproof seal
  • Highly resistant to chemicals and electricity
  • Sterile to prevent sample contamination
These straight-sided cylindrical PP containers are a smart choice for safe sample collection and transportation to laboratories. Ideal for storing diluted bases and acids, and for applications where containers need to keep their shape after bending and flexing. As well as the storage of small electronic parts that need electrical insulation. Sterile and come with a red leakproof PE screw cap.

Containers are protected from particle contamination during shipping. Containers arrive at your site enclosed in a PE bag packed inside a box with a label to show the lot number and expiration date for tracing purposes.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Cap Color
Availability Pricing
Corning Gosselin TP52C-002 Straight-Sided Screw-Cap Container, Red Cap, PP, 125 mL; 380/cs WZ-34551-87 Mfr # TP52C-002
COMPARE
125 Red
Call For Price
Corning Gosselin TP53C-002 Straight-Sided Screw-Cap Container, Red Cap, PP, 180 mL; 264/cs WZ-34551-88 Mfr # TP53C-002
COMPARE
180 Red
Call For Price
Corning Gosselin TP35C-002 Straight-Sided Screw-Cap Container, Red Cap, PP, 60 mL; 700/cs WZ-34551-89 Mfr # TP35C-002
COMPARE
60 Red
Call For Price
Corning Gosselin TP35C-013 Straight-Sided Screw-Cap Container, White Cap, PP, 60 mL; 700/cs WZ-34551-90 Mfr # TP35C-013
COMPARE
60 White
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat