This product is now discontinued, View Related Products
Corning® Gosselin™ PS Seed Germination Culture Dishes
Culture dishes for any application
  • Injection point located near the edge of the bottom maintains optimal clarity for imaging systems Lightweight polystyrene (PS) is compliant with heavy metals (94/62/EC) and food contact (EC 1935/2004) directives Class 100 manufacturing environment ensures aseptic production and packaging Vented for easy gas exchange
Seed germination petri dishes are deeper than traditional dishes (25 mm instead of 15 mm) and feature economical packaging. Choose between dishes with a single chamber or with four chambers. All dishes sterile (SAL 10⁻⁶).
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Sterile
Availability Pricing
Corning Gosselin Seed Germination Petri Dish, 100 x 25 mm, mono; 500/pk WZ-34551-66 Mfr # BT93-02
COMPARE
Optically clear polystyrene, single chamber Yes
Call For Price
Corning Gosselin Seed Germination Petri Dish, 100 x 25 mm, quad; 500/pk WZ-34551-67 Mfr # BTX93-02
COMPARE
Optically clear polystyrene, four chambers Yes