This product is now discontinued, View Related Products
Gloss and Gloss/Thickness Meters
Auto-calibration ensures high accuracy measurements
  • Evaluate the glossy surfaces of coatings, plastics, metals, and paper for quality control applications.
Take this portable gloss meter with you anywhere — meter fits right in your pocket. Display single measurements or statistics including the number of readings, mean, standard deviation, min/max readings, range, difference, and pass/fail. Included easy-link software lets you easily transfer data from the meter to Microsoft Excel® for quality control reports.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Range
Availability Pricing
BYK-Gardner Digital Gloss Meter with USB/Bluetooth Output, 20 Degree; 0 to 2000 GU WZ-59721-41 Mfr # AG-4440
COMPARE
0 to 2000 gloss units
Call For Price
BYK-Gardner Digital Gloss Meter with USB/Bluetooth Output, 60 Degree; 0 to 2000 GU WZ-59721-43 Mfr # AG-4442
COMPARE
0 to 2000 gloss units
Call For Price
BYK-Gardner Digital Gloss Meter with USB/Bluetooth Output, 85 Degree; 0 to 2000 GU WZ-59721-45 Mfr # AG-4444
COMPARE
0 to 2000 gloss units
Call For Price
BYK-Gardner Gloss meter, 20/60/85°, 0 to 2000 GU WZ-59721-47 Mfr # AG-4446
COMPARE
0 to 2000 gloss units at 20°, 0 to 1000 gloss units at 60°, 0 to 160 gloss units at 85°
Call For Price
BYK-Gardner Digital Gloss Meter with USB/Bluetooth Output, 45 Degree; 0 to 180 GU WZ-59721-49 Mfr # AG-4454
COMPARE
0 to 180 gloss units
Call For Price
BYK-Gardner Digital Gloss Meter with USB/Bluetooth Output, 75 Degree; 0 to 140 GU WZ-59721-51 Mfr # AG-4456
COMPARE
0 to 140 gloss units