This product is now discontinued, View Related Products

Bel-Art GHS Labeled Wash Bottles

Eliminate spout dripping with built-in safety vent
  • Quick identification of contents with color-coded caps and chemical identification label
  • Fill bottles easily with wide-mouth design
  • Compliant with OSHA regulations
  • Excellent chemical compatibility with LDPE bottle and polypropylene (PP) cap
These safety-vented wash bottles provide compliance with the Globally Harmonized System (GHS) of Classification and Labeling of Chemicals and OSHA’s HazCom 2012 requirements. These wash bottles are pre-printed with the chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 four-color diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word. These bottles are Deionized Water, Distilled Water, Isotonic Saline, and Water.
These wash bottles feature a unique venting valve which prevents pressure build-up, eliminating potentially dangerous chemical drips. This feature also allows dispensing of the wash bottle in an upright or inverted position. The methyl ethyl ketone and toluene do not include vented valve. Available in a 4-piece assortment pack which includes one each of Acetone, Isopropanol, Bleach, and Ethanol bottles.
Call For Price

22 variations of this product are available.

See All Product Options Below
22 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Cap Color Label Type
Availability Pricing
Bel-Art F12416-0001 GHS Labeled Safety-Vented Acetone Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-60 Mfr # F12416-0001
COMPARE
500 Red Acetone
Call For Price
Bel-Art F12416-0002 GHS Labeled Safety-Vented Dichloromethane Wash Bottles; LDPE, 500 mL; 4/Pk WZ-62300-61 Mfr # F12416-0002
COMPARE
500 Yellow Dichloromethane
Call For Price
Bel-Art F12416-0003 GHS Labeled Safety-Vented Deionized Water Wash Bottles LDPE, 500  mL; 4/Pk WZ-62300-62 Mfr # F12416-0003
COMPARE
500 Blue Deionized Water
Call For Price
Bel-Art F12416-0004 GHS Labeled Safety-Vented Distilled Water Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-63 Mfr # F12416-0004
COMPARE
500 Blue Distilled Water
Call For Price
Bel-Art F12416-0007 GHS Labeled Safety-Vented Ethyl Acetate Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-64 Mfr # F12416-0007
COMPARE
500 Green Ethyl Acetate
Call For Price
Bel-Art F12416-0008 GHS Labeled Safety-Vented Isopropanol Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-65 Mfr # F12416-0008
COMPARE
500 Yellow Isopropanol
Call For Price
Bel-Art F12416-0010 GHS Labeled Safety-Vented Machine Oil Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-66 Mfr # F12416-0010
COMPARE
500 Natural Machine Oil
Call For Price
Bel-Art F12416-0011 GHS Labeled Safety-Vented Methanol Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-67 Mfr # F12416-0011
COMPARE
500 Green Methanol
Call For Price
Bel-Art F12416-0012 GHS Labeled Methyl Ethyl Ketone Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-68 Mfr # F12416-0012
COMPARE
500 Green Methy Ethyl Ketone
Call For Price
Bel-Art F12416-0013 GHS Labeled Safety-Vented Isotonic Saline Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-69 Mfr # F12416-0013
COMPARE
500 Natural Isotonic Saline
Call For Price
Bel-Art F12416-0015 GHS Labeled Safety-Vented Sodium Hypochlorite (Bleach) Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-70 Mfr # F12416-0015
COMPARE
500 Yellow Bleach
Call For Price
Bel-Art F12416-0016 GHS Labeled Toluene Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-71 Mfr # F12416-0016
COMPARE
500 Red Toluene
Call For Price
Bel-Art F12416-0017 GHS Labeled Safety-Vented Water Wash Bottles, LDPE,  500 mL; 4/Pk WZ-62300-72 Mfr # F12416-0017
COMPARE
500 Blue Water
Call For Price
Bel-Art F12416-0019 GHS Labeled Safety-Vented Ethanol Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-73 Mfr # F12416-0019
COMPARE
500 Natural Ethanol
Call For Price
Bel-Art F12416-0050 GHS Labeled Safety-Vented Assorted Wash Bottles, LDPE, 500 mL; 4/Pk WZ-62300-74 Mfr # F12416-0050
COMPARE
500 Red, Yellow, Yellow, and Natural One each of Acetone, Isopropanol, Bleach, and Ethanol
Call For Price
Bel-Art F12432-0001 GHS Labeled Safety-Vented Acetone Wash Bottles, LDPE, 1000 mL; 2/Pk WZ-62300-75 Mfr # F12432-0001
COMPARE
1000 Red Acetone
Call For Price
Bel-Art F12432-0004 GHS Labeled Safety-Vented Distilled Water Wash Bottles, LDPE, 1000 mL; 2/Pk WZ-62300-76 Mfr # F12432-0004
COMPARE
1000 Blue Distilled Water
Call For Price
Bel-Art F12432-0008 GHS Labeled Safety-Vented Isopropanol Wash Bottles, LDPE, 1000 mL; 2/Pk WZ-62300-77 Mfr # F12432-0008
COMPARE
1000 Yellow Isopropanol
Call For Price
Bel-Art F12432-0011 GHS Labeled Safety-Vented Methanol Wash Bottles, LDPE, 1000 mL; 2/Pk WZ-62300-78 Mfr # F12432-0011
COMPARE
1000 Green Methanol
Call For Price
Bel-Art F12432-0015 GHS Labeled Safety-Vented Sodium Hypochlorite (Bleach) Wash Bottles, LDPE, 1000 mL: 2/Pk WZ-62300-79 Mfr # F12432-0015
COMPARE
1000 Yellow Bleach
Call For Price
Showing 1 - 20 of 22

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat