George Fischer Type 546 Ball Valves 42387
Optimize safety with this true union ball valve
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.
£67.17 - £482.14GBP / Each
£80.60 - £578.57GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
GF Signet 161.546.341 Type 546 Ball Valve, PVC with EPDM seals, 3/8" WZ-01357-01 Mfr # 161.546.341
COMPARE
3/8" NPT(F) and socket 3/8" NPT(F) and socket PVC, EPDM
£67.17GBP / Each
£80.60GBP (incl VAT)
GF Signet 161.546.342 Type 546 Ball Valve, PVC with EPDM seals, 1/2" WZ-01357-03 Mfr # 161.546.342
COMPARE
1/2" NPT(F) and socket 1/2" NPT(F) and socket PVC, EPDM
£69.34GBP / Each
£83.21GBP (incl VAT)
GF Signet 161.546.343 Type 546 Ball Valve, PVC with EPDM seals, 3/4" WZ-01357-05 Mfr # 161.546.343
COMPARE
3/4" NPT(F) and socket 3/4" NPT(F) and socket PVC, EPDM
£82.34GBP / Each
£98.81GBP (incl VAT)
GF Signet 161.546.344 Type 546 Ball Valve, PVC with EPDM seals, 1" WZ-01357-07 Mfr # 161.546.344
COMPARE
1" NPT(F) and socket 1" NPT(F) and socket PVC, EPDM
£99.68GBP / Each
£119.62GBP (incl VAT)
GF Signet 163.546.351 Type 546 Ball Valve, CPVC with seals, 3/8" WZ-01371-08 Mfr # 163.546.351
COMPARE
3/8" NPT(F) and socket 3/8" NPT(F) and socket CPVC, Viton®
£123.52GBP / Each
£148.22GBP (incl VAT)
GF Signet 163.546.352 Type 546 Ball Valve, CPVC with seals, 1/2" WZ-01371-09 Mfr # 163.546.352
COMPARE
1/2" NPT(F) and socket 1/2" NPT(F) and socket CPVC, Viton®
£125.68GBP / Each
£150.82GBP (incl VAT)
GF Signet 163.546.353 Type 546 Ball Valve, CPVC with seals, 3/4" WZ-01371-10 Mfr # 163.546.353
COMPARE
3/4" NPT(F) and socket 3/4" NPT(F) and socket CPVC, Viton®
£153.85GBP / Each
£184.62GBP (incl VAT)
GF Signet TYPE-546 Type 546 Ball Valve, Manual, PP, 1/2"NPT(F), FPM Seals WZ-01374-92 Mfr # 175.546.332(TYPE-546)
COMPARE
1/2" NPT(F) 1/2" NPT(F) PVDF, PTFE, FPM
£316.37GBP / Each
£379.64GBP (incl VAT)
GF Signet TYPE-546 Type 546 Ball Valve, Manual, PP, 1"NPT(F), FPM Seals WZ-01374-94 Mfr # 175.546.334(TYPE-546)
COMPARE
1" NPT(F) 1" NPT(F) PVDF, PTFE, FPM
£482.14GBP / Each
£578.57GBP (incl VAT)
Showing 1- 9 of 9