General-Purpose Watertight Single Setpoint Switches
Trigger alarms or shut down equipment when pressure rises to setpoint
General-Purpose Watertight Dual Set Point Switches are ideal for use with pumps, paint spraying equipment, liquid level, and presses. Use these watertight switches in breweries, dairies, chemical plants, and pulp/paper mills.
Housing meets NEMA 4X standards. Epoxy-coated aluminum enclosure mounts with integral wall mounting bracket with two additional knock-out plugs for installation. SS internal parts. Remove cover to read scale while adjusting set points.
£204.32 - £325.00GBP / Each
£245.18 - £390.00GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Ashcroft B4200PSI 0 to 200 psi General-Purpose Watertight Switch, Single Setpoint WZ-68050-13 Mfr # B4-24-B-XCY-200#
COMPARE
200
£204.32GBP / Each
£245.18GBP (incl VAT)
Ashcroft 0 to 215 psi General-Purpose Pressure Switch WZ-98077-02 Mfr # B421T-XFS-400PSI (SET @ 215PSI RISING)
COMPARE
215
£325.00GBP / Each
£390.00GBP (incl VAT)