General-Purpose TEFC/ODP, NEMA Type C-Face Four-In-One Motors
Totally enclosed motors are ideal for dusty, dirty, or damp environments
  • Unique four-in-one design allows for replacement of ODP, TEFC, C-Face, or rigid based motors
  • Use for vertical or horizontal applications
Single-phase motors include a removable, rigid base. Three-phase motors inlcude a removable base.
£321.15 - £536.18GBP / Each
£385.38 - £643.42GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Motor RPM (rpm) Motor Frame Size Motor hp
Availability Pricing
COMPARE
3600 56C 1/2
£321.15GBP / Each
£385.38GBP (incl VAT)
COMPARE
1800 56C 1/2
£420.97GBP / Each
£505.16GBP (incl VAT)
COMPARE
1800 56C 3/4
£443.01GBP / Each
£531.61GBP (incl VAT)
COMPARE
1800 56HC 1
£536.18GBP / Each
£643.42GBP (incl VAT)
COMPARE
1800 56C 1/3
£389.24GBP / Each
£467.09GBP (incl VAT)
COMPARE
3600 56C 1/2
£330.88GBP / Each
£397.06GBP (incl VAT)
COMPARE
1800 56C 1/2
£427.93GBP / Each
£513.52GBP (incl VAT)
COMPARE
3600 56C 3/4
£371.48GBP / Each
£445.78GBP (incl VAT)
COMPARE
1800 56C 3/4
£446.00GBP / Each
£535.20GBP (incl VAT)
Showing 1- 9 of 9