Veolia Z.Plex Depth Cartridge Filters
100% greater dirt-holding capacity lowers total cost of filter operations
  • Up to twice the life of conventional depth filters
  • Up to 50% lower pressure drop
Z.Plex® filters are made from thermally bonded fibers of polypropylene. GE Water & Process Technologies certifies that it uses no resin binders, lubricants, antistatic or release agents, or other additives in the manufacture of these cartridges. Suitable to 160°F (71°C).
ROSave.Z® filters
For use in reverse osmosis pretreatment and other pure water applications. Available in 1 and 5 micron pore sizes. The resin used for manufacturing the filter media meets the food contact requirement of the U.S., and is FDA compliant.
Applications
• Bottled water
• Beverage
• Electronics
WellPro.Z® filters
For use in well injection, produced water disposal and other oil and gas applications. Available in 1, 5, 10 and 20 micron pore sizes.
Applications
• Well injection
• Produced water filtration
• Water flood / enhanced oil recovery
• Brine filtration
• Sea water filtration
£10.29 - £22.05GBP / Each
£12.35 - £26.46GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
Veolia 1236581 Depth CartridFilter, 5.0 micron, plain end, 10" WZ-29831-61 Mfr # RO.ZS 05-10/1236581
COMPARE
10 6 GPM at 1.0 psid 5
£10.29GBP / Each
£12.35GBP (incl VAT)
Veolia 1236577 Depth CartridFilter, 1.0 micron, plain end, 10" WZ-29831-63 Mfr # RO.ZS 01-10/1236577
COMPARE
10 3.2 GPM at 1.0 psid 1
£11.60GBP / Each
£13.92GBP (incl VAT)
Veolia 1236263 Depth CartridFilter, 5.0 micron, plain end, 20" length WZ-29831-65 Mfr # RO.ZS 05-20/1236263
COMPARE
20 12 GPM at 1.0 psid 5
£18.90GBP / Each
£22.68GBP (incl VAT)
Veolia 1236262 Depth CartridFilter, 1.0 micron, plain end, 20" length WZ-29831-67 Mfr # RO.ZS 01-20/1236262
COMPARE
20 6.4 GPM at 1.0 psid 1
£22.05GBP / Each
£26.46GBP (incl VAT)
Veolia 1238938 Depth CartridFilter, 10 micron, plain end, 20" length WZ-29831-69 Mfr # WP.ZS 10-20/1238938
COMPARE
20 5 GPM at 0.35 psid 10
£11.16GBP / Each
£13.39GBP (incl VAT)
Veolia 1238937 Depth CartridFilter, 5 micron, plain end, 20" length WZ-29831-73 Mfr # WP.ZS 05-20/1238937
COMPARE
20 5 GPM at 0.92 psid 5
£10.99GBP / Each
£13.19GBP (incl VAT)
Veolia 1238936 Depth CartridFilter, 1 micron, plain end, 20" length WZ-29831-75 Mfr # WP.ZS 01-20/1238936
COMPARE
20 5 GPM at 0.45 psid 1
£12.69GBP / Each
£15.23GBP (incl VAT)
Showing 1- 7 of 7