This product is now discontinued, View Related Products
GasAlertExtreme Single-Gas Detectors
Replaceable sensor and batteries extend life of detector
  • Easy four-button operation
  • Lithium battery provides up to 9000 hours of use
The easy-to-read LCD shows continuous real-time gas concentrations and instrument status. The detectors feature simple auto zero and calibration procedures. All models feature a 90 dB alarm and bright LED to warn you of the presence of hazardous conditions.
The LOW, HIGH, and TWA alarms have adjustable set points; peak values can be displayed on demand. The full function self-test verifies sensor, circuit, and battery integrity and activates the audible/visual/vibrating alarms each time you turn on the instrument.
Approval: UL Class I, Div. 1, Groups A, B, C, D; CE
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
BW Technologies GAXTHD Single-Gas Detector, Hydrogen Sulfide, with data logging WZ-10105-09 Mfr # GAXTHD
COMPARE
H2S
Call For Price
BW Technologies GAXTMD Extreme Single-Gas Detector, Carbon Monoxide, with data logging WZ-10105-10 Mfr # GAXTMD
COMPARE
Call For Price
BW Technologies GAXTSD Single-Gas Detector, Sulfur Dioxide, with data logging WZ-10105-11 Mfr # GAXTSD
COMPARE
SO2
Call For Price
BW Technologies GAXTXD Single-Gas Detector, Oxygen, with data logging WZ-10105-12 Mfr # GAXT-X-DL-2
COMPARE
O2
Call For Price
BW Technologies GAXT-M Single-Gas Detector, Carbon Monoxide WZ-81991-11 Mfr # GAXT-M-DL
COMPARE
CO
Call For Price
BW Technologies GAXT-H Extreme Single-Gas Detector, Hydrogen Sulfide WZ-81991-13 Mfr # GAXT-H
COMPARE
Call For Price
BW Technologies GAXT-X Extreme Single-Gas Detector, Oxygen WZ-81991-15 Mfr # GAXT-X-DL-2
COMPARE
Call For Price
BW Technologies GAXT-S Extreme Single-Gas Detector, Sulfur Dioxide WZ-81991-17 Mfr # GAXT-S
COMPARE
Call For Price
Showing 1- 8 of 8