Labconco FreeZone Freeze Dryer Manifolds
Manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Accommodates ½” and ¾” adapters for quick connection of flasks
Stainless steel manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks.
£1,168.00 - £3,809.09GBP / Each
£1,401.60 - £4,570.91GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Labconco FreeZone Freeze Drying Manifold, 20 Ports; FreeZone FD WZ-03348-31 Mfr # 7522300
COMPARE
Freeze Drying Manifold, 20 Ports; FreeZone FD
£3,809.09GBP / Each
£4,570.91GBP (incl VAT)
Labconco FreeZone Freeze Drying Manifold, 4 Ports; FreeZone FD WZ-03353-11 Mfr # 7201700
COMPARE
Freeze Drying Manifold, 4 Ports; FreeZone FD
£1,168.00GBP / Each
£1,401.60GBP (incl VAT)
Labconco FreeZone Freeze Drying Manifold, 8 Ports; FreeZone FD WZ-03353-12 Mfr # 7201800
COMPARE
Freeze Drying Manifold, 8 Ports; FreeZone FD
£2,526.68GBP / Each
£3,032.02GBP (incl VAT)