This product is now discontinued, View Related Products

Gillis Mfg Four-shelf Vented Units

Corrosion resistance with 1000-lb capacity per shelf!
  • Protect and organize your products with these high-density structural foam polyethylene shelvingunits
Each shelf features a 1"-square steel tubular frame with baked plastic resin coating. Rugged 16-gauge steel posts ensure stability and also have the plastic coating. Shelves are available in solid or vented construction. Vented units
provide excellent visibility and allow the air to move freely. Shelves feature a lifetime guarantee against rust.
Customize your shelving units by ordering the starter set, and then attaching add-on units to extend your shelves, or to turn a 90° corner. The add-on units have legs on one end only, the other connects at the shelves of the starter units.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Number of Shelves Description
Availability Pricing
Gillis Mfg PS1836V72S-4 Starter four-shelf vented units, 36"L x 18"W WZ-86411-20 Mfr # PS1836V72S-4
COMPARE
1000 4 Starter four-shelf vented units, 36\"L x 18\"W
Call For Price
Gillis Mfg PS1848V72S-4 Starter four-shelf vented units, 48"L x 18"W WZ-86411-21 Mfr # PS1848V72S-4
COMPARE
1000 4 Starter four-shelf vented units, 48\"L x 18\"W
Call For Price
Gillis Mfg PS1860V72S-4 Starter four-shelf vented units, 60"L x 18"W WZ-86411-22 Mfr # PS1860V72S-4
COMPARE
1000 4 Starter four-shelf vented units, 60\"L x 18\"W
Call For Price
Gillis Mfg PS2436V72S-4 Starter four-shelf vented units, 36"L x 24"W WZ-86411-23 Mfr # PS2436V72S-4
COMPARE
1000 4 Starter four-shelf vented units, 36\"L x 24\"W
Call For Price
Gillis Mfg PS2448V72S-4 Starter four-shelf vented units, 48"L x 24"W WZ-86411-24 Mfr # PS2448V72S-4
COMPARE
1000 4 Starter four-shelf vented units, 48\"L x 24\"W
Call For Price
Gillis Mfg PS2460V72S-4 Starter four-shelf vented units, 60"L x 24"W WZ-86411-25 Mfr # PS2460V72S-4
COMPARE
1000 4 Starter four-shelf vented units, 60\"L x 24\"W
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat