This product is now discontinued, View Related Products

Gillis Mfg Four-shelf Solid Units

Corrosion resistance with 1000-lb capacity per shelf!
  • Protect and organize your products with these high-density structural foam polyethylene shelving units
Each shelf features a 1"-square steel tubular frame with baked plastic resin coating. Rugged 16-gauge steel posts ensure stability and also have the plastic coating. Shelves are available in solid or vented construction. Solid shelves are accepted by the USDA for direct food contact. Shelves feature a lifetime guarantee against rust.
Customize your shelving units by ordering the starter set, and then attaching add-on units to extend your shelves, or to turn a 90° corner. The add-on units have legs on one end only, the other connects at the shelves of the starter units.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Shelf Length (in) Shelf Width (in)
Availability Pricing
Gillis Mfg PS1836S72S-4 Starter four-shelf solid unit, 36"L x 18"W WZ-86411-00 Mfr # PS1836S72S-4
COMPARE
1000 18 36
Call For Price
Gillis Mfg PS1848S72S-4 Starter four-shelf solid unit, 48"L x 18"W WZ-86411-01 Mfr # PS1848S72S-4
COMPARE
1000 18 48
Call For Price
Gillis Mfg PS1860S72S-4 Starter four-shelf solid unit, 60"L x 18"W WZ-86411-02 Mfr # PS1860S72S-4
COMPARE
1000 18 60
Call For Price
Gillis Mfg PS2436S72S-4 Starter four-shelf solid unit, 36"L x 24"W WZ-86411-03 Mfr # PS2436S72S-4
COMPARE
1000 24 36
Call For Price
Gillis Mfg PS2448S72S-4 Starter four-shelf solid unit, 48"L x 24"W WZ-86411-04 Mfr # PS2448S72S-4
COMPARE
1000 24 48
Call For Price
Gillis Mfg PS2460S72S-4 Starter four-shelf solid unit, 60"L x 24"W WZ-86411-05 Mfr # PS2460S72S-4
COMPARE
1000 24 60
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat