This product is now discontinued, View Related Products
Fluke 2638A/05 Hydra Series III 22-Channel Data Acquisition System and DMM
Precision data collection in a portable stand-alone system
  • High-performance system accepts up to 22 inputs to measure voltage, current, resistance, temperature, and more
  • Outstanding DC voltage accuracy of ±0.0024%!
  • Features a dedicated 6.5-digit DMM
  • Supports 15 common thermocouple types
  • Data transfer to PC via USB memory stick or remote USB/LAN
  • Set dual alarm set points for each channel to easily monitor data
  • 20 on-board math channels with alarm settings for complex calculations
  • 6 TTL Alarm outputs assignable to one or multiple channels
Excellent accuracy, full-color display, easy-to-use menu system, and industrial safety ratings make the Hydra series III data acquisition system a standout in its class. This 22-channel (20 universal inputs and 2 dedicated low-burden current inputs) data logger with universal input module accepts AC/DC voltage, AC/DC current, resistance, frequency, thermocouple, RTD, and thermistor inputs.
The Hydra series III system features a removable universal input module that plugs into a rear slot to give you the flexibility to leave your module at the test site, completely wired, and ready to use. When you need data from that site, simply "plug in" and load your saved setup file. The calibration date and input module serial number are programmed into the module and are read by the data logger when installed—ideal for collecting data in a regulated industry.
A dedicated 6.5-digit multimeter provides additional functionality and value. On-screen color trend graphing and analysis offer multichannel real-time data display. Simply push a button to capture the chart results or data displays for reports or a closer review. Statistics include rate of change, Min, Max, standard deviation, average, sample size, and peak-to-peak information. A 20 MB on-board memory stores up to 75,000 data records. A built-in USB port collects and stores files up to 32 GB directly to a USB flash drive.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Voltage (VAC) Voltage Accuracy (VAC) Description
Availability Pricing
Fluke Calibration 2638A/05 Hydra Series III 22-Channel Data Acquisition System and DMM WZ-16108-25 Mfr # 2638A/05 120
COMPARE
Hydra Series III 22-Channel Data Acquisition System and DMM
Call For Price
Fluke Calibration 2638A/05/C Hydra Series III 22-Channel Data Acquisition System WZ-16108-27 Mfr # 2638A/05/C 120
COMPARE
Hydra Series III 22-Channel Data Acquisition System