This product is now discontinued, View Related Products
Fluke 1750 Three-Phase Power Recorders
Never miss hard-to-find issues while monitoring critical equipment
  • Eliminate costly downtime by capturing power quality events to uncover equipment malfunction
  • Automatically store complete power system data on the memory card or export via the Ethernet connection
  • Monitor all measurements on every cycle with outstanding accuracy and resolution
  • Set up in minutes without a PC and with self-identifying current probes and single-lead voltage connections
These Fluke 1750 three-phase power recorders allow you to easily and accurately measure and validate voltage and current on three phases, neutral, and ground of critical equipment. All measurements comply with IEC61000-4-30 standards for correct evaluation of all measured values.
Safely capture power quality events without needing batteries or external power supplies. These recorders only require single-lead voltage connections which enhances safety while also allowing quick setup. Certified to meet the strictest safety standards, these comply for use in CAT IV-600 V and CAT III-1000 V environments.
Every cycle on every channel is simultaneously sampled to provide you with a complete analysis of power quality, harmonics, power and energy. Cross-channel current and voltage triggering will capture event data on all input channels, no matter which channel triggered the event. This way you will never miss capturing a disturbance again.
The three models that include the Windows® 10 tablet allow analysis to be performed in the field with the use of the Fluke Power Analyze software. Voice, data, and picture annotations can be made to flag important points on the tablet as you are recording measurements. The easy-to-use interface lets you to view data the way you want, and even modify thresholds after data is recorded.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
FLUKE-1750-TF/NT Three-Phase Power Recorder with four 1,000A flexible current probes without PC tablet WZ-20003-00 Mfr # FLUKE-1750-TF/NT
COMPARE
Three-Phase Power Recorder with four 1,000A flexible current probes without PC tablet
Call For Price
FLUKE-1750/NT Three-Phase Power Recorder with 400A current probes without PC tablet WZ-20003-01 Mfr # FLUKE-1750/NT
COMPARE
Three-Phase Power Recorder with 400A current probes without PC tablet
Call For Price
FLUKE-1750/B/NT Three-Phase Power Recorder without current probes or PC tablet WZ-20003-02 Mfr # FLUKE-1750/B/NT
COMPARE
Three-Phase Power Recorder without current probes or PC tablet
Call For Price
FLUKE-1750-TF/ET Three-Phase Power Recorder with four 1,000A flexible current probes and PC tablet WZ-20003-03 Mfr # FLUKE-1750-TF/ET
COMPARE
Three-Phase Power Recorder with four 1,000A flexible current probes and PC tablet
Call For Price
FLUKE-1750/ET Three-Phase Power Recorder with four 400A current probes and PC tablet WZ-20003-04 Mfr # FLUKE-1750/ET
COMPARE
Three-Phase Power Recorder with four 400A current probes and PC tablet
Call For Price
FLUKE-1750/B/ET Three-Phase Power Recorder without current probes with PC tablet WZ-20003-05 Mfr # FLUKE-1750/B/ET
COMPARE
Three-Phase Power Recorder without current probes with PC tablet