Flowline EchoSafe Explosion-Proof Ultrasonic Level transmitter (2-wire)
  • Compact XP enclosure with viewing window and dual conduit ports
  • Narrow 3'' beam diameter enables flexible installation in applications with limited measurement space
  • Rugged corrosion resistant PVDF transducer and process mount for chemical and wastewater
  • Fail-safe intelligence with diagnostic feedback for easy troubleshooting
  • Display indicates air gap or level height distance in inch or centimeter
EchoSafe™, an explosion proof, two-wire ultrasonic transmitter provides noncontact level measurement up to 10m in hazardous bulk storage, day tank and sump applications, and is well suited for challenging corrosive, slurry or waste media. Media examples include sodium hypochlorite, wastewater and resin. The integral display and push button module enable fast and simple calibration.
£1,237.40 - £1,421.40GBP / Each
£1,484.88 - £1,705.68GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
Flowline XP88-0 Explosion-Proof Ultrasonic Level Transmitter WZ-68873-81 Mfr # XP88-0
COMPARE
24.6 30
£1,237.40GBP / Each
£1,484.88GBP (incl VAT)
Flowline XP89-0 Explosion-Proof Ultrasonic Level transmitter (2-wire), 32 8ft WZ-68873-82 Mfr # XP89-0
COMPARE
32.8 30
£1,421.40GBP / Each
£1,705.68GBP (incl VAT)