Flowline DeltaSpan Submersible Pressure Level Transmitters
Accurate and corrosion resistant
  • Corrosion resistant 316 SS transducers with polyurethane or ETFE cables
  • Strong cable design with maintenance free breather tube and vent filter
  • Automatic temperature compensation for accurate measurement
£708.40 - £1,150.00GBP / Each
£850.08 - £1,380.00GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
Flowline SD31-S101 Submersible Pressure Level Transmitter WZ-68046-41 Mfr # LD31-S101
COMPARE
£708.40GBP / Each
£850.08GBP (incl VAT)
Flowline LD32-S101 Submersible Pressure Level Transmitter, slurry WZ-68046-43 Mfr # LD32-S101
COMPARE
£1,062.60GBP / Each
£1,275.12GBP (incl VAT)
Flowline LD31-S301 Intrinsically Safe Pressure Level Transmitter WZ-68046-45 Mfr # LD31-S301
COMPARE
£828.00GBP / Each
£993.60GBP (incl VAT)
Flowline LD32-S301 Intrinsically Safe Pressure Level Transmitter WZ-68046-47 Mfr # LD32-S301
COMPARE
£1,150.00GBP / Each
£1,380.00GBP (incl VAT)