Flow Ratchet Tubing Control Clamps
Won't rust, corrode, or distort
  • Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
£18.88 - £757.16GBP / Pkg of 250
£22.66 - £908.59GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Housing Material Pinch Valve Tube Size
Availability Pricing
COMPARE
Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack Polyoxymethylene (POM) Acetal 1/8" to 1/2" OD
£49.84GBP / Pkg of 25
£59.81GBP (incl VAT)
COMPARE
Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/Pk Polyoxymethylene (POM) Acetal 1/16" to 1/4" OD
£18.88GBP / Pkg of 25
£22.66GBP (incl VAT)
COMPARE
Ratchet Tubing Clamp for up to 1/2" Tubing, Acetalp; 250/Pk Polyoxymethylene (POM) Acetal 1/8" to 1/2" OD
£757.16GBP / Pkg of 250
£908.59GBP (incl VAT)
COMPARE
Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 250/Pk Polyoxymethylene (POM) Acetal 1/16" to 1/4" OD
£198.83GBP / Pkg of 250
£238.60GBP (incl VAT)