This product is now discontinued, View Related Products
Flir T420bx, T440bx Advanced Building Thermal Imagers
  • UltraMax gives you 4X the resolution
  • MSX Multi-Spectral Dynamic Imaging.
  • The industries leading high resolution cameras with scalable Fusion (PIP) and a touch-screen Keypad that has been optimized to take advantage of the unique FLIR user interface
  • Highest accuracy for its class at 2%
  • Copy images and reports to USB flash drive
  • Instant in-camera multipage report generation
  • Thermal fusion blends thermal and digital images in real time
  • Bluetooth™ transmitter with METERLiNK® wireless communication
  • Auto-orientation function
The T420 and T440 infrared cameras arm users with superior infrared imaging resolution that works harder to reveal trouble spots. With sharp thermal resolution at 76,800 pixels (320x240) for accurate diagnostics even from a distance. Thermal imaging is a non-contact technology that measures infrared wavelengths to determine temperatures from a safe distance. A thermal imager displays an image using different colors to represent different temperatures.
With the highest accuracy for its class at 2% and thermal sensitivity as low as 0.045°C, these imagers are perfect for identifying hot spots or a rise in temperature which often indicates a problem. FLIR's thermal imagers are fully radiometric which means they measure and store temperatures at every point in the image. Each camera comes with access to robust web based software package that allows the user to transfer images onto a template for analyzing and creating of a report in pdf format. The T-series also includes built-in software which allows complete in-field analysis, reporting and file sharing capability directly from the camera. FLIR-Report a professional thermal imaging software package is sold separately and provides enhanced reporting capabilities, allowing for the easy creating of comprehensive multi-image reports with analysis of image data.
MSX™ Multi-Spectral Dynamic Imaging, a FLIR exclusive. MSX adds the detail of real-time visible spectrum images captured by the built-in digital camera to thermal spectrum images, providing extraordinary sharpness, contrast and clarity previously unavailable in thermal imaging.
UltraMax gives you 4X the resolution and combined with FLIR’s exclusive MSX Technology it substantially enhances IR images when imported into FLIR Tools software. UltraMax resolution: 307,200 pixels (640x480.
For building-industry professionals, “bx” versions quickly alert users to moisture intrusion and insulation problems with humidity and insulation alarms.
METERLiNK® is a Bluetooth™ connection that is an exclusive feature of the FLIR E-Series cameras and select Extech instruments. Measurements from these instruments are transmitted and stored directly on the thermal image and displayed on the camera’s viewing screen. Allowing the user a quick way to review and record multiple parameters at the location being inspected.
FLIR Tools Mobile app streams video and connects FLIR imagers to Apple® and Android™ devices for improved client communication
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Detector Size Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Flir T420bx Building Thermal Imaging Camera; MSx/25 Degree Lens SKU 3975415 WZ-39754-15 Mfr # FLIR T420BX
COMPARE
320 x 240 662 76,800
Call For Price
Flir T420bx Building Thermal Imaging Camera; MSx/25 Degree Lens SKU 3975417 WZ-39754-17 Mfr # FLIR T440BX
COMPARE
320 x 240 662 76,800