This product is now discontinued, View Related Products

FLIR® IRW-Series Thermal Inspection Windows

Helps reduce the vast majority of dangerous arc flash occurrences
  • Rugged frame handles both outdoor and indoor use
  • Decrease downtime and increase efficiency with easy installation
  • Easy flip-open hatch for faster inspections
  • Simple thumb screw releases the permanently hinged cover
Opening electrical cabinets to perform thermal and visual inspections of live components is dangerous work, exposing you to the risk of a hazardous arc flash incident. The FLIR IRW-series of thermal inspection windows help eliminate that danger. The window allows all brands of thermal imagers to capture both visible light and thermal images along with allowing LED and laser illumination to pass through the glass. A secure, permanently hinged cover opens easily with one hand, which means there’s nothing to remove, drop, or lose. The IRW-Series windows are comprehensively tested for safety and have a limited lifetime warranty.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Width (in) Description
Availability Pricing
Flir IRW-SS-2C Infrared Inspection Window, Stainless Steel, 2" WZ-39756-32 Mfr # IRW-SS-2C
COMPARE
260 2 7/8 Infrared Inspection Window, Stainless Steel, 2\"
Call For Price
Flir IRW-SS-3C Infrared Inspection Window, Stainless Steel, 3" WZ-39756-33 Mfr # IRW-SS-3C
COMPARE
260 3 57/64 Infrared Inspection Window, Stainless Steel, 3\"
Call For Price
Flir IRW-SS-4C Infrared Inspection Window, Stainless Steel, 4" WZ-39756-34 Mfr # IRW-SS-4C
COMPARE
260 5 Infrared Inspection Window, Stainless Steel, 4\"
Call For Price
Flir IRW-6PC Infrared Inspection Window, Aluminum, 6" WZ-39758-03 Mfr # IRW-6PC
COMPARE
200 6 19/64 Infrared Inspection Window, Aluminum, 6\"
Call For Price
Flir IRW-6PS Infrared Inspection Window, Powder-Coated Stainless Steel, 6" WZ-39758-06 Mfr # IRW-6PS
COMPARE
200 6 19/64 Infrared Inspection Window, Powder-Coated Stainless Steel, 6\"
Call For Price
Flir IRW-12PS Infrared Inspection Window, Powder-Coated Stainless Steel, 12" WZ-39758-07 Mfr # IRW-12PS
COMPARE
200 12 Infrared Inspection Window, Powder-Coated Stainless Steel, 12\"
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat