This product is now discontinued, View Related Products
Flir Exx-Series Advanced Thermal Imagers with MSX (E53, E75, E85 and E95)
Superior resolution and range performance needed to quickly discover potential points of failure
  • Vibrant, 4” optically-bonded rapid-response touch screen with intuitive new user interface
  • Convenient menu buttons allow for one-handed operation
  • Unique folder and naming structure that makes finding images easier
  • Connect over Wi-Fi to mobile devices or via METERLiNK® to FLIR instruments
  • Rugged design with IP54 rated for demanding applications
  • FLIR’s industry-leading 2-5-10 warranty
FLIR® Exx-series thermal imagers are extremely lightweight, compact and rugged. Thermal imagers are fully radiometric. They measure and store temperatures at every point in the image. Fusion technology provides enhanced problem detection and analysis capabilities through the blending of visual and infrared images. The built-in laser pointer also helps associate the physical target with the hot spot on the thermal image, indicating where a problem exists.
The vibrant, 4” screen features the latest Dark Precision touch screen interface at your fingertips; you’ll be able to instantly tap into multiple temperature measurement tools, image modes, archived JPEGs, and more. Auto screen orientation lets you switch between landscape and portrait view to optimize your field of view while keeping the temperature measurement overlays upright on the LCD no matter how you hold the camera. Designed for continuous operation on a single charge for up to 2.5 hours, the unit features a rechargeable Li-ion battery with AC adapter/charger.
Flir’s exclusive Multi-Spectral Dynamic Imaging System, known as MSX®, is included on all E-series cameras which adds visible spectrum definition to thermal images in real time for amazing detail. This function provides extraordinary thermal detail that instantly highlights and orients problem locations and eliminates the need to refer back to a visual image. MSX allows you to see details never available before in earlier generations of thermal imagers, from definable edges of an object to text on a label and so much more.
All E-Series cameras feature Wi-Fi connections; simply download the FLIR Viewer app to your Apple® or Android™ mobile device and link to your thermal imager. Then transfer images to your mobile device for further analysis—also great for readjusting span, level, or palettes, creating reports, and e-mailing results. Optional FLIR-Report professional thermal imaging software package enhances reporting capabilities and lets you easily create comprehensive multi-image reports with analysis of image data.
METERLiNK™ is a Bluetooth® connection that is an exclusive feature on FLIR Exx-Series cameras. Measurements from METERLiNK instruments (sold separately) are transmitted and stored directly onto the thermal imagers and displayed on the camera’s viewing screen, allowing the user a quick way to review and record multiple parameters at the location being inspected.
All thermal imager features
• ±3.6°F (±2°C) or ±2% of reading for ambient temperature
• 4" Color 640 x 480 optically bonded touch screen with selectable on-screen viewing modes
• 5-megapixel digital visible light camera with lamp
• Picture-in-picture to overlay thermal and digital images
• Compass with GPS image tagging
• Adjustable emissivity
• Replaceable Li-ion rechargeable battery
• Isotherm and automatic hot/cold detection marker
• Alarms modes include moisture, insulation, and measurements
• 1 to 4x digital zoom
• Streaming video
• Voice, text, and sketch annotation
• MPEG 4 video recording
• METERLiNK via Bluetooth®
• Wi-Fi app connectivity
Call For Price
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Pixels / Thermal Resolution Description
Availability Pricing
Flir E53 Thermal Imaging Camera with MSX, 240 x 180 Resolution (43,200 Pixels) and 24° Lens (Standard) WZ-39756-83 Mfr # FLIR E53
COMPARE
650 43200 Thermal Imaging Camera with MSX, 240 x 180 Resolution (43,200 Pixels) and 24° Lens (Standard)
Call For Price
Flir E75 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 14° Lens WZ-39756-85 Mfr # FLIR E75-14
COMPARE
650 76800 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 14° Lens
Call For Price
Flir E85 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 14° Lens WZ-39756-86 Mfr # FLIR E85-14
COMPARE
1200 110592 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 14° Lens
Call For Price
Flir E95 Thermal Imaging Camera with MSX, 3464 x 348 Resolution (161,472 Pixels) and 14° Lens WZ-39756-87 Mfr # FLIR E95-14
COMPARE
1500 161472 Thermal Imaging Camera with MSX, 3464 x 348 Resolution (161,472 Pixels) and 14° Lens
Call For Price
Flir E75 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° Lens (Standard) WZ-39757-75 Mfr # Flir E75-24
COMPARE
650 76800 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° Lens (Standard)
Call For Price
Flir E75 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 42° Lens WZ-39757-76 Mfr # Flir E75-42
COMPARE
650 76800 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 42° Lens
Call For Price
Flir E75 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° + 42° Lens WZ-39757-77 Mfr # Flir E75-24-42
COMPARE
650 76800 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° + 42° Lens
Call For Price
Flir E75 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° + 14° Lens WZ-39757-78 Mfr # Flir E75-24-14
COMPARE
650 76800 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° + 14° Lens
Call For Price
Flir E75 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° + 42° + 14° Lens WZ-39757-79 Mfr # FLIR E75241442
COMPARE
650 76800 Thermal Imaging Camera with MSX, 320 x 240 Resolution (76,800 Pixels) and 24° + 42° + 14° Lens
Call For Price
Flir E85 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° Lens (Standard) WZ-39757-85 Mfr # Flir E85-24
COMPARE
1200 110592 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° Lens (Standard)
Call For Price
Flir E85 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 42° Lens WZ-39757-86 Mfr # Flir E85-42
COMPARE
1200 110592 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 42° Lens
Call For Price
Flir E85 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° + 42° Lens WZ-39757-87 Mfr # Flir E85-24-42
COMPARE
1200 110592 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° + 42° Lens
Call For Price
Flir E85 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° + 14° Lens WZ-39757-88 Mfr # Flir E85-24-14
COMPARE
1200 110592 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° + 14° Lens
Call For Price
Flir E85 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° + 42° + 14° Lens WZ-39757-89 Mfr # FLIR E85241442
COMPARE
1200 110592 Thermal Imaging Camera with MSX, 384 x 288 Resolution (81,792 Pixels) and 24° + 42° + 14° Lens
Call For Price
Flir E95 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° Lens (Standard) WZ-39757-95 Mfr # Flir E95-24
COMPARE
1500 161472 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° Lens (Standard)
Call For Price
Flir E95 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 42° Lens WZ-39757-96 Mfr # Flir E95-42
COMPARE
1500 161472 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 42° Lens
Call For Price
Flir E95 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° + 42° Lens WZ-39757-97 Mfr # Flir E95-24-42
COMPARE
1500 161472 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° + 42° Lens
Call For Price
Flir E95 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° + 14° Lens WZ-39757-98 Mfr # Flir E95-24-14
COMPARE
1500 161472 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° + 14° Lens
Call For Price
Flir E95 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° + 42° + 14° Lens WZ-39757-99 Mfr # FLIR E95241442
COMPARE
1500 161472 Thermal Imaging Camera with MSX, 464 x 348 Resolution (161,472 Pixels) and 24° + 42° + 14° Lens
Call For Price
Showing 1- 19 of 19