Flare to NPT(M) threaded straight adapters
Leak-tight and high-pressure connection for corrosive chemical applications
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.
£50.61 - £128.93GBP / Each
£60.73 - £154.72GBP (incl VAT)
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
Parker Hannifin Flare compression pipe adapter, 1/4" OD x 1/8" NPT(M), PFA, Each WZ-30490-18 Mfr # FAMS-42
COMPARE
1/4 PFA 1/8
£50.61GBP / Each
£60.73GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 1/4" OD x 1/4" NPT(M), PFA, Each WZ-30490-20 Mfr # FAMS-44
COMPARE
1/4 PFA 1/4
£50.61GBP / Each
£60.73GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 1/4" OD x 3/8" NPT(M), PFA, Each WZ-30490-22 Mfr # FAMS-46
COMPARE
1/4 PFA 3/8
£50.61GBP / Each
£60.73GBP (incl VAT)
Parker Hannifin FAMS-48 Flare compression pipe adapter, 1/4" OD x 1/2" NPT(M), PFA, Each WZ-30490-24 Mfr # FAMS-48
COMPARE
1/4 PFA 1/2
£60.67GBP / Each
£72.80GBP (incl VAT)
Parker Hannifin FAMS-62 Flare compression pipe adapter, 3/8" OD x 1/8" NPT(M), PFA, Each WZ-30490-26 Mfr # FAMS-62
COMPARE
3/8 PFA 1/8
£56.34GBP / Each
£67.61GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 3/8" OD x 1/4" NPT(M), PFA, Each WZ-30490-28 Mfr # FAMS-64
COMPARE
3/8 PFA 1/4
£56.34GBP / Each
£67.61GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 3/8" OD x 3/8" NPT(M), PFA, Each WZ-30490-30 Mfr # FAMS-66
COMPARE
3/8 PFA 3/8
£63.92GBP / Each
£76.70GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 3/8" OD x 1/2" NPT(M), PFA, Each WZ-30490-32 Mfr # FAMS-68
COMPARE
3/8 PFA 1/2
£66.09GBP / Each
£79.31GBP (incl VAT)
Parker Hannifin FAMS-82 Flare compression pipe adapter, 1/2" OD x 1/8" NPT(M), PFA, Each WZ-30490-34 Mfr # FAMS-82
COMPARE
1/2 PFA 1/8
£67.17GBP / Each
£80.60GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 1/2" OD x 1/4" NPT(M), PFA, Each WZ-30490-36 Mfr # FAMS-84-P
COMPARE
1/2 PFA 1/4
£102.93GBP / Each
£123.52GBP (incl VAT)
Parker Hannifin FAMS-86-P Flare compression pipe adapter, 1/2" OD x 3/8" NPT(M), PFA, Each WZ-30490-38 Mfr # FAMS-86-P
COMPARE
1/2 PFA 3/8
£102.93GBP / Each
£123.52GBP (incl VAT)
Parker Hannifin Flare compression pipe adapter, 1/2" OD x 1/2" NPT(M), PFA, Each WZ-30490-40 Mfr # FAMS-88
COMPARE
1/2 PFA 1/2
£70.43GBP / Each
£84.52GBP (incl VAT)
Parker Hannifin FAMS-128 Flare compression pipe adapter, 3/4" OD x 1/2" NPT(M), PFA, Each WZ-30490-42 Mfr # FAMS-128
COMPARE
3/4 PFA 1/2
£100.76GBP / Each
£120.91GBP (incl VAT)
Parker Hannifin FAMS-1212-P Flare compression pipe adapter, 3/4" OD x 3/4" NPT(M), PFA, Each WZ-30490-44 Mfr # FAMS-1212-P
COMPARE
3/4 PFA 3/4
£128.93GBP / Each
£154.72GBP (incl VAT)
Showing 1- 14 of 14