Fixing Clamps for Domnick Hunter Air Purification Systems
Optimize performance of your Parker® domnick hunter Air Purification Systems
Fixing/Mounting Clamps join adjacent filters mounted in series. Clamp also functions as a wall-mounting bracket to minimize associated hardware. These clamps are compatible with 29587-series filters.
£96.00 - £108.00GBP / Each
£115.20 - £129.60GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Parker Hannifin FXKE1 Fixing Clamp for 21 cfm Domnick Hunter Air Filters WZ-29587-80 Mfr # FXKE1
COMPARE
Fixing Clamp for 21 cfm Domnick Hunter Air Filters
£96.00GBP / Each
£115.20GBP (incl VAT)
Parker Hannifin FXKE2 Fixing Clamp For 42 And 64 cfm Filters WZ-29587-82 Mfr # FXKE2
COMPARE
Fixing Clamp For 42 And 64 cfm Filters
Call For Price
Parker Hannifin FXKE3 Fixing Clamp For 127 And 233 cfm Filters WZ-29587-84 Mfr # FXKE3
COMPARE
Fixing Clamp For 127 And 233 cfm Filters
£108.00GBP / Each
£129.60GBP (incl VAT)