This product is now discontinued, View Related Products

Filters for 30-Inch Fume Hoods

Protect yourself from hazardous fumes, vapors, odors, particles, and contaminants with one of four types of filters for 30” self-monitoring ductless fume hoods
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Exterior Width (in) Filter Type
Availability Pricing
Primary Hydrocarbon Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-50 Mfr # AU-2804
COMPARE
30 Primary Hydrocarbon
Call For Price
Secondary Hydrocarbon Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-55 Mfr # AU-5504
COMPARE
30 Secondary Hydrocarbon
Call For Price
Primary Mineral Acid Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-60 Mfr # AU-2814
COMPARE
42 Primary Mineral Acid
Call For Price
Secondary Mineral Acid Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-65 Mfr # AU-5214
COMPARE
30
Call For Price
Primary Formaldehyde Filters for 30" Self-Monitoring Ductless Fume Hoods (2 required) WZ-33713-70 Mfr # AU-2807
COMPARE
42 Primary Formaldehyde
Call For Price
Secondary Formaldehyde Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-75 Mfr # AU-5207
COMPARE
30 Secondary Formaldehyde
Call For Price
Primary HEPA Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-80 Mfr # AU-2820
COMPARE
42 Primary HEPA
Call For Price
Secondary HEPA Filters for 30" Self-Monitoring Ductless Fume Hoods WZ-33713-85 Mfr # AU-5220
COMPARE
30 Secondary HEPA
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat