BriskHeat® Fibrox High-Temperature Heating Tapes leads opposite ends-120 VAC
Flexible, multipurpose, direct-contact heaters are ideal for use on conductive surfaces
  • Maximum exposure temperature of 900°F (482°C)
  • Choose from power leads on same end or on opposite ends
  • Heavy-duty insulated design
  • Nongrounded
These high-temperature heating tapes are made of resistance wires that are insulated with braided fiberglass knitted into a flat tape and covered with heavy fiberglass fabric.
The 120 VAC tapes with an opposite-end lead configuration are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC two-prong plug.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
£113.01 - £510.75GBP / Each
£135.61 - £612.90GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat TBIH 051-100 Fibrox Insulated Heating Tape Leads On Opposite Ends, 10' x 1/2" , 520 Watts, 120 V WZ-03105-10 Mfr # TBIH 051-100
COMPARE
10 900 120
£197.28GBP / Each
£236.74GBP (incl VAT)
BriskHeat TBIH 051-020 Fibrox Insulated Heating Tape Leads On Opposite Ends, 2' x 1/2" , 104 Watts, 120 V WZ-03105-20 Mfr # TBIH 051-020
COMPARE
2 900 120
£120.03GBP / Each
£144.04GBP (incl VAT)
BriskHeat TBIH 051-040 Fibrox Insulated Heating Tape Leads On Opposite Ends, 4' x 1/2" , 208 Watts, 120 V WZ-03105-40 Mfr # TBIH 051-040
COMPARE
4 900 120
£127.69GBP / Each
£153.23GBP (incl VAT)
BriskHeat TBIH 051-060 Fibrox Insulated Heating Tape Leads On Opposite Ends, 6' x 1/2" , 312 Watts, 120 V WZ-03105-60 Mfr # TBIH 051-060
COMPARE
6 900 120
£134.71GBP / Each
£161.65GBP (incl VAT)
BriskHeat TBIH 051-080 Fibrox Insulated Heating Tape Leads On Opposite Ends, 8' x 1/2" , 416 Watts, 120 V WZ-03105-80 Mfr # TBIH 051-080
COMPARE
8 900 120
£179.83GBP / Each
£215.80GBP (incl VAT)
BriskHeat TBIH 101-020 Fibrox Insulated Heating Tape Leads On Opposite Ends, 2' x 1" , 208 Watts, 120 V WZ-03106-20 Mfr # TBIH 101-020
COMPARE
2 900 120
£113.01GBP / Each
£135.61GBP (incl VAT)
BriskHeat TBIH-101-040 Fibrox Insulated Heating Tape Leads On Opposite Ends, 4' x 1" , 416 Watts, 120 V WZ-03106-40 Mfr # TBIH-101-040
COMPARE
4 900 120
£184.09GBP / Each
£220.91GBP (incl VAT)
BriskHeat TBIH-101-060 Fibrox Insulated Heating Tape Leads On Opposite Ends, 6' x 1" , 624 Watts, 120 V WZ-03106-60 Mfr # TBIH-101-060
COMPARE
6 900 120
£233.67GBP / Each
£280.40GBP (incl VAT)
BriskHeat TBIH-171-100 Fibrox Insulated Heating Tape Leads On Opposite Ends, 10' x 1-3/4" , 1040 Watts WZ-03107-10 Mfr # TBIH-171-100
COMPARE
10 900 120
£510.75GBP / Each
£612.90GBP (incl VAT)
BriskHeat TBIH-171-020 Fibrox Insulated Heating Tape Leads On Opposite Ends, 2' x 1-3/4" , 208 Watts, 120 V WZ-03107-20 Mfr # TBIH-171-020
COMPARE
2 900 120
£209.41GBP / Each
£251.29GBP (incl VAT)
BriskHeat TBIH-171-060 Fibrox Insulated Heating Tape Leads On Opposite Ends, 6' x 1-3/4" , 624 Watts, 120 V WZ-03107-60 Mfr # TBIH-171-060
COMPARE
6 900 120
£381.36GBP / Each
£457.63GBP (incl VAT)
Showing 1- 11 of 11