This product is now discontinued, View Related Products
Norwell Dipstik Reusable Bailers, FEP
Sink fast for quicker sampling
  • Available in two styles a nd lengths
  • Order polyester suspension cord separately
FEP Reusable Bailers are available in two styles and lengths. Economy bailers have a 1-3/4" OD and feature a fixed top and bottom ball valve. Standard bailers have a 1-7/8" OD and feature interchangeable top and bottom cap configurations. Standard bailers come in six of the most popular top/bottom cap configurations.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description Length (ft) Material
Availability Pricing
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X1', with standard top/bottom WZ-05497-00 Mfr # D8653000
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X1', with standard top/bottom 1 FEP, PTFE
Call For Price
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X3', with standard top/bottom WZ-05497-05 Mfr # D8653005
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X3', with standard top/bottom 3 FEP, PTFE
Call For Price
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X1', with standard top/emptying bottom WZ-05497-10 Mfr # D8653020
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X1', with standard top/emptying bottom 1 FEP, PTFE
Call For Price
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X3', with standard top/emptying bottom WZ-05497-15 Mfr # D8653025
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X3', with standard top/emptying bottom 3 FEP, PTFE
Call For Price
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X1', with standard top/flow control bottom WZ-05497-20 Mfr # D8653050
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X1', with standard top/flow control bottom 1 FEP, PTFE
Call For Price
Norwell Dipstik Reusable, FEP bailer, 1-7/8"OD X1', with point source top/standard bottom WZ-05497-30 Mfr # D8653030
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X1', with point source top/standard bottom 1 FEP, PTFE
Call For Price
Norwell Dipstik D8653040 Reusable, FEP bailer, 1-7/8"OD X1', with point source top/emptying bottom WZ-05497-40 Mfr # D8653040
COMPARE
Reusable, FEP bailer, 1-7/8\"OD X1', with point source top/emptying bottom FEP, PTFE
Call For Price
Norwell Dipstik Economy, reusable, FEP bailer, 1-7/8"OD X1', with fixed top/ball bottom WZ-05497-60 Mfr # D8653070
COMPARE
Economy, reusable, FEP bailer, 1-7/8\"OD X1', with fixed top/ball bottom 1 FEP, PTFE
Call For Price
Norwell Dipstik Economy, reusable, FEP bailer, 1-7/8"OD X3', with fixed top/ball bottom WZ-05497-65 Mfr # D8653075
COMPARE
Economy, reusable, FEP bailer, 1-7/8\"OD X3', with fixed top/ball bottom 3 FEP, PTFE
Call For Price
Showing 1- 9 of 9