This product is now discontinued, View Related Products
External Thread Slotted Skirt Polypropylene Cryogenic Vials
Color coded caps enable quick sample identification
Vials are free standing with a conical bottom and feature 0.5 ml graduations. Vials are designed for use in the vapor phase of liquid nitrogen and are ideal for storage or transport of material at extremely low temperatures. External threads provide a smooth inner surface. The screw cap incorporates a molded-in sealing ring for a tight, leak proof seal.
Slotted Skirt Cryogenic Vials are freezable to -40°C with molded graduations and feature multiple cap colors. Choose sterile or non-sterile for a wide variety of applications.
Call For Price
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
Compareitem specifications
Cap color Tube Bottom Shape Volume (mL)
Availability Pricing
COMPARE
Assorted Slotted skirt 2
Call For Price
Qorpak AKM-3100-0006 External Thread PP Cryogenic Vials, Blue/Slot/2.0 mL; 1000/Cs WZ-03756-64 Mfr # AKM-3100-0006
COMPARE
Blue Slotted skirt 2
Call For Price
Qorpak AKM-3100-0005 External Thread PP Cryogenic Vials, Sterile/Blue/Slot/2.0 mL; 500/Cs WZ-03756-65 Mfr # AKM-3100-0005
COMPARE
Blue Slotted skirt 2
Call For Price
COMPARE
Green Slotted skirt 2
Call For Price
Qorpak AKM-3100-0011 External Thread PP Cryogenic Vials, Sterile/Green/Slot/2.0 mL; 500/Cs WZ-03756-67 Mfr # AKM-3100-0011
COMPARE
Green Slotted skirt 2
Call For Price
COMPARE
Orange Slotted skirt 2
Call For Price
COMPARE
Orange Slotted skirt 2
Call For Price
COMPARE
Red Slotted skirt 2
Call For Price
COMPARE
Red Slotted skirt 2
Call For Price
COMPARE
White Slotted skirt 2
Call For Price
COMPARE
White Slotted skirt 2
Call For Price
COMPARE
Yellow Slotted skirt 2
Call For Price
COMPARE
Yellow Slotted skirt 2
Call For Price
COMPARE
Blue Slotted skirt 2.5
Call For Price
COMPARE
Green Slotted skirt 2.5
Call For Price
COMPARE
Orange Slotted skirt 2.5
Call For Price
COMPARE
Red Slotted skirt 2.5
Call For Price
COMPARE
White Slotted skirt 2.5
Call For Price
COMPARE
Yellow Slotted skirt 2.5
Call For Price
COMPARE
Blue Slotted skirt 3.5
Call For Price
Showing 1 - 20 of 29