Extech Waterproof Palm pH Meter
Big Digit LCD display in rugged housing with choice of stick or cabled pH Electrode
  • Large LCD displays pH or mV and temperature simultaneously
  • Microprocessor provides for automatic buffer recognition
  • Memory stores 15 readings
  • Automatic temperature compensation via Pt-100 sensor built into electrode
  • Unit is completely waterproof when electrode is connected to the meter
  • Rugged design for handheld or wall mount
  • Magnetic back panel offers optimum flexibility and convenience
  • Low battery indicator and auto power off
Specifically designed for fast response measurement.
£195.00GBP / Each
£234.00GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max pH (pH) Min pH (pH) Description
Availability Pricing
Extech PH220-C Palm pH Meter with Cabled Electrode WZ-53026-43 Mfr # PH220-C
COMPARE
14 0 Palm pH Meter with Cabled Electrode
£195.00GBP / Each
£234.00GBP (incl VAT)