This product is now discontinued, View Related Products
Extech MultiMaster Precision Multimeters
Multimeters designed for your professional needs
  • Rugged housing protects meter in demanding applications
These CAT IV, 600V meters are made to withstand the most industrial applications. MultiMaster™ meters (26866-00 & -10) are AC/DC True-RMS, high accuracy multimeters. Features include trigger level, auto-off, continuity test, relative, hold, and peak capture. For process measurements, a 4 to 20 mA signal displays as 0 to 100%. Download data to a PC via RS-232 input (order cable and software 26866-20). Model 26866-10 also measures temperature.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Extech MM560 Precision High Accuracy Multimeter WZ-26866-00 Mfr # MM560A
COMPARE
1000
Call For Price
Extech MM570 Precision Multimeter with Temperature WZ-26866-10 Mfr # MM570A
COMPARE
1000