Extech Desktop Indoor Air Quality Carbon Dioxide Monitor
Monitor air quality in schools, office buildings, greenhouses, factories, and anywhere that carbon dioxide is a concern
  • Measures and displays carbon dioxide concentration, air temperature and humidity
  • Captures min/max; press RESET to clear memory
  • Displays the year, month, date, and time of day
  • Audible alarm sounds when carbon dioxide levels exceed the set point
  • Activates a
Monitor air quality in schools, office buildings, greenhouses, factories, and anywhere that carbon dioxide is a concern. Measures and displays carbon dioxide concentration, air temperature and humidity. Captures min/max; press RESET to clear memory. Also displays the year, month, date, and time of day. Audible alarm sounds when carbon dioxide levels exceed the set point. Activates an alarm relay that can be used for connection to an external indicating or controlling device.
£342.50GBP / Each
£411.00GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max CO2 (ppm) Max Temperature (° F) Output
Availability Pricing
Extech CO210 Desktop Indoor Air Quality COMonitor with Memory WZ-81973-71 Mfr # CO210
COMPARE
9999 140 USB
£342.50GBP / Each
£411.00GBP (incl VAT)