Extech Data Logging Pressure Meter with SD Card
Records data on an SD card in Excel® format
  • Accepts interchangeable transducers with ranges of 30, 150, 300 psi (transducers sold separately)
  • Meter automatically senses plug-in transducer''s range and calibration
  • Displays 10 types of pressure units
  • Offset adjustment used for zero function to make relative measurements
  • Adjustable data sampling rate: 1 second to 8 hrs, 59 min, 59 sec
  • Stores 99 readings manually and 20M readings via 2G SD card
  • Large backlit LCD
  • Record/recall Min/Max, Data Hold, auto power-off
£370.20GBP / Each
£444.24GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Data Storage (samples) Description
Availability Pricing
Extech SDL700 Pressure Meter with SD Card Datalogging in Excel format WZ-68602-28 Mfr # SDL700
COMPARE
99 internal, 2 GB (SD card) Pressure Meter with SD Card Datalogging in Excel format
£370.20GBP / Each
£444.24GBP (incl VAT)