Extech Current and Voltage Calibrator
Light, compact design is ideal for field use
  • Provides adjustable 0 to 24 mA and 0 to 10 V calibration source
  • Five preset calibration values enable faster calibrations
Use this calibrator to set up and perform calibrations of process devices such as transducers, indicators, recorders, controllers and more. Dual-function calibrator simulates (source mode) and displays (measure mode) current and voltage outputs. Unit offers high accuracy (0.025%) with 24 VDC that drives current loads up to 1000 Ω. The “flip-up” design is easy-to-use and the six-button keypad makes calibration set ups and tests quick and simple. The large LCD displays readings in mA, as a percentage of the 0 to 24 mA range (4 to 20 mA = 100.0%), or as mV in voltage ranges.
£304.55GBP / Each
£365.46GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Measurement (mA) Max Measurement (VDC)
Availability Pricing
Extech 412355A Current & Voltage Calibrator, 110 to 240 VAC WZ-16120-10 Mfr # 412355A
COMPARE
£304.55GBP / Each
£365.46GBP (incl VAT)