This product is now discontinued, View Related Products
Extech 700 Series 800A Clamp Meters
Functions include Inrush, Capacitance and Frequency
FEATURES
  • Models with AC/DC Current, AC/DC Voltage, Resistance, Frequency, Capacitance, Temperature, Diode and Continuity
  • 1ms Fast Peak Hold captures Inrush currents and transients
  • True RMS models
  • Double molded housing for better grip and one-handed operation
  • Large LCD display with bright backlighting
  • Data hold "freezes" data in display
  • 1.2" (30mm) circular jaw opening for large cables (500MCM)
  • Auto power off
  • Complete with test leads, 9V battery and Type K bead wire probe (EH720, EH730) and carrying case
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Extech EX730 AC/DC TRMS Clamp Meter, 800 Amps WZ-20003-20 Mfr # EX730
COMPARE
Call For Price
Extech EX710 800 Amp AC Current Clamp Meter WZ-20005-36 Mfr # EX710
COMPARE
Call For Price
Extech EX720 True RMS AC Clamp Meter, 800 Amps WZ-20005-37 Mfr # EX720