Extech 600A True RMS Calmp Meters + Noncontact Voltage
Stay safe—check for live wires first
  • Built-in non-contact Voltage detector with LED alert
  • Multimeter functions include AC current, AC/DC voltage, resistance, capacitance, frequency, temperature, duty cycle, diode, and continuity
  • Relative mode for capacitance zero and offset adjustment
  • Data Hold, Min/Max readings, and auto power-off
£67.00 - £200.00GBP / Each
£80.40 - £240.00GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacitance Max (mF) Resistance Max (MΩ) Description
Availability Pricing
Extech MA610 600A Clamp Meter + Non Contact Voltage WZ-20000-89 Mfr # MA610
COMPARE
0.1 40 Clamp Meter Non Contact Voltage
£67.00GBP / Each
£80.40GBP (incl VAT)
Extech MA620-K Digital Multimeter/Clamp Meter Test Kit WZ-20034-54 Mfr # MA620-K
COMPARE
4 60 Digital Multimeter/Clamp Meter Test Kit
£190.00GBP / Each
£228.00GBP (incl VAT)
Extech MA640-K Phase Rotation/Clamp Meter Test Kit WZ-20034-55 Mfr # MA640-K
COMPARE
4 60 Phase Rotation/Clamp Meter Test Kit
£200.00GBP / Each
£240.00GBP (incl VAT)
Extech MA620 600A, True RMS AC Clamp Meter + Non Contact Voltage WZ-26823-30 Mfr # MA620
COMPARE
4 60 True RMS AC Clamp Meter Non Contact Voltage
£124.00GBP / Each
£148.80GBP (incl VAT)
Extech MA640 600A, True RMS AC/DC Clamp Meter + Non Contact Voltage WZ-26823-32 Mfr # MA640
COMPARE
4 60 True RMS AC/DC Clamp Meter Non Contact Voltage
£152.00GBP / Each
£182.40GBP (incl VAT)