Extech 400 Series Multimeters
Precision accuracy at an economical price
  • True RMS AC voltage and AC current readings
  • IR Thermometer with 8:1 distance to target ratio
  • Audible and visual input alerts
  • Frequency and capacitance capabilities
  • Low current capability - measure down to 0.1 µA
  • MAX (Model 450) function
  • Easy access battery door
  • Backlight Display
  • IEC CAT III Safety Rating
£51.55GBP / Each
£61.86GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Extech EX410 DMM with Temperature (Type K) 0.5% Accuracy WZ-20005-31 Mfr # EX410A
COMPARE
£51.55GBP / Each
£61.86GBP (incl VAT)