BEVCO ESD Vinyl Cleanroom Chairs
Ideal for use in semiconductor, bioscience, pharmaceutical, and board manufacturing
  • 300 lb capacity
All chairs feature heavy-duty construction, easy-to-use pneumatic height adjustment, and 360° swivel. Each chair has a sturdy five-legged base with glides; order optional casters separately.
These ESD/ISO 6 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12.1-2013 delivering a combination ESD/Cleanroom seating solution for highly technical environments.
£420.94 - £594.60GBP / Each
£505.13 - £713.52GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Seat height (in) Description
Availability Pricing
Bevco 4050E3-BLK Desk Height ESD/ISO 6 Cleanroom Black Vinyl Chair; AL Base WZ-86309-09 Mfr # 4050E3-BLK
COMPARE
ESD Vinyl 15.5 to 20.5 Westmound Desk Height ESD/ISO 6 Cleanroom Black Vinyl Chair; Non-Tilt; Polished Aluminum Base
£420.94GBP / Each
£505.13GBP (incl VAT)
Bevco 4550E3-BLK Tall Height ESD/ISO 6 Cleanroom Black Vinyl Chair; AL Base w/Adj. Footring WZ-86309-15 Mfr # 4550E3-BLK
COMPARE
ESD Vinyl 23 to 33 Westmound Tall Height ESD/ISO 6 Cleanroom Black Vinyl Chair; Polished Aluminum Base w/Adjustable Footring
£491.17GBP / Each
£589.40GBP (incl VAT)
Bevco 9050ME4-BLK Desk Height ESD/ISO 7 Cleanroom Black Vinyl Chair; Medium Back; AL Base WZ-86309-40 Mfr # 9050ME4-BLK
COMPARE
ESD Vinyl 15.5 to 21 Integra Desk Height ESD/ISO 7 Cleanroom Black Vinyl Chair; Medium Back; Non-Tilt; Polished Aluminum Base
£515.86GBP / Each
£619.03GBP (incl VAT)
Bevco 9050ME1-BLK Desk Height ESD/ISO 4 Cleanroom Black Vinyl Chair; Medium Back; AL Base WZ-86309-42 Mfr # 9050ME1-BLK
COMPARE
ESD Vinyl 15.5 to 21 Integra Desk Height ESD/ISO 4 Cleanroom Black Vinyl Chair; Medium Back; Non-Tilt; Polished Aluminum Base
£589.49GBP / Each
£707.39GBP (incl VAT)
Bevco 9350ME4-BLK Mid Height ESD/ISO 7 Cleanroom Black Vinyl Chair; Medium Back; AL Base w/Adj. Footring WZ-86309-43 Mfr # 9350ME4-BLK
COMPARE
ESD Vinyl 19 to 26.5 Integra Mid Height ESD/ISO 7 Cleanroom Black Vinyl Chair; Medium Back; Polished Aluminum Base w/Adjustable Footring
£594.60GBP / Each
£713.52GBP (incl VAT)