This product is now discontinued, View Related Products
Environmental Express Trace Metal Standard Air Filters
Added quality control to your industrial hygiene testing
  • Meets NIOSH Method 7300 requirements
  • Industrial hygiene standards for trace metals
Filters are 37-mm diameter mixed cellulose ester (MCE) with a 0.8 µm pore size. The concentrations are reported as µg per filter, and are certified to ±1%. Expiration date of one year.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Trace Metals Filters, 1 to 10 µg, 37 mm; 10 filters, 5 blanks
Call For Price
COMPARE
Trace Metals Filters, 10 to 50 µg, 37 mm; 10 filters, 5 blanks
Call For Price
COMPARE
7300 Trace Metals Filters, 10 to 100 µg, 37 mm; 10 filters, 5 blanks
Call For Price
COMPARE
Trace Metals Filters, 0.1-50ug/37mm; 10 filters, 5 blanks
Call For Price
COMPARE
Trace Metals Filters, 0.2 to 100 µg, 37 mm; 10 filters, 5 blanks
Call For Price
COMPARE
Trace Metals Filters, 1 to 10 µg, 37 mm; 10 filters, 5 blanks
Call For Price
COMPARE
Trace Metals Filters, 5 to 50 µg, 37 mm; 10 filters, 5 blanks
Call For Price
Showing 1- 7 of 7