Environmental Express® Filter Membranes
Prevent sample-to-sample contamination
  • Resistant to strong acids and bases
Ideal replacement membrane filters for your radiological extration filter funnels and SimpleDist micro tubes.
£93.70 - £230.60GBP / Pkg of 1
£112.44 - £276.72GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
25 Polypropylene 0.1
£93.70GBP / Pkg of 1
£112.44GBP (incl VAT)
COMPARE
47 Polypropylene 0.1
£153.50GBP / Pkg of 1
£184.20GBP (incl VAT)
COMPARE
£230.60GBP / Pkg of 50
£276.72GBP (incl VAT)