This product is now discontinued, View Related Products
Elma Elmasonic E+ Ultrasonic Cleaners
Protect sensitive items—safety shutoff and temperature-selectable alarm light
  • Easily preselect run time and temperature with rotary dials
  • Obtain uniform sound waves and cleaning from the sweep function
  • Activate the pulse function for 20% increase in ultrasonic power and soil removal
The Elmasonic E+ series of ultrasonic cleaners feature simple control with superior cleaning technology. Select heating temperature from 30 to 80°C, in 5° increments, with a rotary dial. Choose a maximum temperature for sensitive items and instruments—LED light illuminates if reached. Heating not available on model 59988-14. Set to operate continuously or in timed runs, from 1 to 30 minutes, also selected with a dial.
The integrated sweep mode ensures even sound distribution throughout the bath at 37 kHz for efficient and effective cleaning. Turn on pulse mode to increase ultrasonic power 20%—ideal for those hard to remove soils and pastes. Safety features include auto-shutoff if running for 8 hours continuously or if bath temperature exceeds 90°C, and a sealed display and elevated feet to protect against splashes and spills. Baths with capacities above 1 gallon also feature a side-mounted drain for easy emptying and cleaning.
Call For Price
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Frequency Output (KHz) Power (VAC)
Availability Pricing
Elmasonic E+ EP10H Ultrasonic Cleaner with Heat, 0.25 gal.; 120 VAC WZ-59988-13 Mfr # 107 1647
COMPARE
0.2 37 120
Call For Price
Elmasonic E+ EP10  Ultrasonic Cleaner, 0.25 gal.; 120 VAC WZ-59988-14 Mfr # 107 1649
COMPARE
0.2 37 120
Call For Price
Elmasonic E+ EP20H Ultrasonic Cleaner with Heat, 0.5 gal.; 120 VAC WZ-59988-15 Mfr # 107 1653
COMPARE
0.5 37 120
Call For Price
Elmasonic E+ EP30H Ultrasonic Cleaner with Heat, 0.75 gal.; 120 VAC WZ-59988-16 Mfr # 107 1657
COMPARE
0.7 37 120
Call For Price
Elmasonic E+ EP40H Ultrasonic Cleaner with Heat, 1.25 gal.; 120 VAC WZ-59988-17 Mfr # 107 1665
COMPARE
1.1 37 120
Call For Price
Elmasonic E+ EP60H Ultrasonic Cleaner with Heat, 1.5 gal.; 120 VAC WZ-59988-18 Mfr # 107 1667
COMPARE
1.5 37 120
Call For Price
Elmasonic E+ EP100H Ultrasonic Cleaner with Heat, 2.5 gal.; 120 VAC WZ-59988-19 Mfr # 107 1670
COMPARE
2.5 37 120
Call For Price
Elmasonic E+ EP120H Ultrasonic Cleaner with Heat, 3.5 gal.; 120 VAC WZ-59988-20 Mfr # 107 1672
COMPARE
3.4 37 120
Call For Price
Elmasonic E+ EP180H Ultrasonic Cleaner with Heat, 4.75 gal.; 120 VAC WZ-59988-21 Mfr # 107 1674
COMPARE
4.75 37 120
Call For Price
Elmasonic E+ EP300H Ultrasonic Cleaner with Heat, 7.5 gal.; 120 VAC WZ-59988-22 Mfr # 107 1676
COMPARE
7.4 37 120
Call For Price
Elmasonic E+ EP10H Ultrasonic Cleaner with Heat, 0.25 gal.; 220 VAC WZ-59989-13 Mfr # 107 1646-CP
COMPARE
0.2 37 220
Call For Price
Elmasonic E+ EP10  Ultrasonic Cleaner, 0.25 gal.; 220 VAC WZ-59989-14 Mfr # 107 1648-CP
COMPARE
0.2 37 220
Call For Price
Elmasonic E+ EP20H Ultrasonic Cleaner with Heat, 0.5 gal.; 220 VAC WZ-59989-15 Mfr # 107 1652-CP
COMPARE
0.5 37 220
Call For Price
Elmasonic E+ EP30H Ultrasonic Cleaner with Heat, 0.75 gal.; 220 VAC WZ-59989-16 Mfr # 107 1656-CP
COMPARE
0.7 37 220
Call For Price
Elmasonic E+ EP40H Ultrasonic Cleaner with Heat, 1.25 gal.; 220 VAC WZ-59989-17 Mfr # 107 1664-CP
COMPARE
1.1 37 220
Call For Price
Elmasonic E+ EP60H Ultrasonic Cleaner with Heat, 1.5 gal.; 220 VAC WZ-59989-18 Mfr # 107 1666-CP
COMPARE
1.5 37 220
Call For Price
Elmasonic E+ EP100H Ultrasonic Cleaner with Heat, 2.5 gal.; 220 VAC WZ-59989-19 Mfr # 107 1669-CP
COMPARE
2.5 37 220
Call For Price
Elmasonic E+ EP120H Ultrasonic Cleaner with Heat, 3.5 gal.; 220 VAC WZ-59989-20 Mfr # 107 1671-CP
COMPARE
3.4 37 220
Call For Price
Elmasonic E+ EP180H Ultrasonic Cleaner with Heat, 4.75 gal.; 220 VAC WZ-59989-21 Mfr # 107 1673-CP
COMPARE
4.75 37 220
Call For Price
Elmasonic E+ EP300H Ultrasonic Cleaner with Heat, 7.5 gal.; 220 VAC WZ-59989-22 Mfr # 107 1675-CP
COMPARE
7.4 37 220
Call For Price
Showing 1- 20 of 20